EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
A protocol for an asymmetric synthesis of γ-amino acids.
PMID 22747507
Abstract
A new and practical method for the asymmetric synthesis of γ-amino acids from β,γ-butenolides by an in situ esterification, condensation, and reduction in a one-pot procedure is described. This method is quite general for the preparation of both enantiomers of aryl or aliphatic γ-amino acids in high yields. These γ-amino-acid derivatives were also shown to be versatile synthetic intermediates for further transformations by their conversion to γ-lactams, δ-amino alcohols, and hydrolysis products in high yields with no racemization.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
283754
2(5H)-Furanone, 98%
C4H4O2