EMAIL THIS PAGE TO A FRIEND
Journal of fish biology
Cardiac remodelling of Atlantic halibut Hippoglossus hippoglossus induced by experimental anaemia with phenylhydrazine.
PMID 22747824
Abstract
Phenylhydrazine injections (0.3 mg kg(-1) , followed by a second injection of 0.1 mg kg(-1) 7 days later) induced a reproducible and stable anaemia in Atlantic halibut Hippoglossus hippoglossus, reducing the haematocrit and haemoglobin by 70.0 and 75.5%, respectively, over 3 weeks. There were no changes in blood electrolyte or lactate concentrations, although anaemic fish showed a 37.5 and 33.0% increase in cardiac somatic index and ventricular somatic index, respectively, compared with dimethyl sulphur oxide (DMSO) and saline vehicle controls. Changes in cardiac somatic indices did not correlate with the ratio of ventricular length:height and length:width did correlate with haematocrit and haemoglobin indicating that changes in cardiac shape may occur as a function of anaemic hypoxemia.