EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Efficient synthesis and 5-LOX/COX-inhibitory activity of some 3-hydroxybenzo[b]thiophene-2-carboxylic acid derivatives.
PMID 22749420
Abstract
A series of 3-hydroxybenzo[b]thiophene-2-carboxylic acid derivatives has been prepared and subsequently evaluated with regards to the inhibition of 5-LOX/COX. Structure optimization furnished derivatives with promising in vitro activity as dual 5-LOX/COX inhibitors with submicromolar IC(50) values for inhibition of 5-LOX and COX-1, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T32603
2-Thiophenecarboxylic acid, ReagentPlus®, 99%
C5H4O2S