EMAIL THIS PAGE TO A FRIEND
Journal of child neurology
Vigabatrin and mental retardation in tuberous sclerosis: infantile spasms versus focal seizures.
PMID 22752486
Abstract
Tuberous sclerosis complex is a genetic disorder resulting in epilepsy and mental retardation. Vigabatrin has shown efficacy in the treatment of infantile spasms caused by tuberous sclerosis complex, but its effects on focal seizures caused by tuberous sclerosis complex have not been determined. We compared the efficacy of vigabatrin in patients with tuberous sclerosis complex-induced focal seizures and infantile spasms and assessed the mental outcomes in both groups. We retrospectively evaluated 31 children with tuberous sclerosis complex and epilepsy, who were treated with vigabatrin in a single tertiary center in Seoul, Korea. Vigabatrin treatment resulted in spasms cessation in 16 of 18 (88.9%) patients with infantile spasms, whereas 6 of 13 (46.2%) patients with focal seizures became seizure free. Initial response to vigabatrin had no effect on intellectual disability. Vigabatrin was highly effective in eliminating infantile spasms caused by tuberous sclerosis complex but was less effective in patients with focal seizures.
Related Materials