EMAIL THIS PAGE TO A FRIEND
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America
Current state of Clostridium difficile treatment options.
PMID 22752868
Abstract
Recent reports of reduced response to standard therapies for Clostridium difficile infection (CDI) and the risk for recurrent CDI that is common with all currently available treatment agents have posed a significant challenge to clinicians. Current recommendations include metronidazole for treatment of mild to moderate CDI and vancomycin for severe CDI. Results from small clinical trials suggest that nitazoxanide and teicoplanin may be alternative options to standard therapies, whereas rifaximin has demonstrated success in uncontrolled trials for the management of multiple recurrences. Anecdotal reports have also suggested that tigecycline might be useful as an adjunctive agent for the treatment of severe complicated CDI. Reports of resistance will likely limit the clinical use of fusidic acid and bacitracin and, possibly, rifaximin if resistance to this agent becomes widespread. Treatment of patients with multiple CDI recurrences and those with severe complicated CDI is based on limited clinical evidence, and new treatments or strategies are needed.