EMAIL THIS PAGE TO A FRIEND
Journal of microbiology (Seoul, Korea)
Paenibacillus xylaniclasticus sp. nov., a xylanolytic-cellulolytic bacterium isolated from sludge in an anaerobic digester.
PMID 22752902
Abstract
A mesophilic, facultative, anaerobic, xylanolytic-cellulolytic bacterium, TW1(T), was isolated from sludge in an anaerobic digester fed with pineapple waste. Cells stained Gram-positive, were spore-forming, and had the morphology of straight to slightly curved rods. Growth was observed in the temperature range of 30 to 50°C (optimum 37°C) and the pH range of 6.0 to 7.5 (optimum pH 7.0) under aerobic and anaerobic conditions. The strain contained meso-diaminopimelic acid in the cell-wall peptidoglycan. The predominant isoprenoid quinone was menaquinone with seven isoprene units (MK-7). Anteiso-C(15:0), iso-C(16:0), anteiso-C(17:0), and C(16:0) were the predominant cellular fatty acids. The G+C content of the DNA was 49.5 mol%. A phylogenetic analysis based on 16S rRNA showed that strain TW1(T) belonged within the genus Paenibacillus and was closely related to Paenibacillus cellulosilyticus LMG 22232(T), P. curdlanolyticus KCTC 3759(T), and P. kobensis KCTC 3761(T) with 97.7, 97.5, and 97.3% sequence similarity, respectively. The DNA-DNA hybridization values between the isolate and type strains of P. cellulosilyticus LMG 22232(T), P. curdlanolyticus KCTC 3759(T), and P. kobensis KCTC 3761(T) were found to be 18.6, 18.3, and 18.0%, respectively. The protein and xylanase patterns of strain TW1(T) were quite different from those of the type strains of closely related Paenibacillus species. On the basis of DNA-DNA relatedness and phenotypic analyses, phylogenetic data and the enzymatic pattern presented in this study, strain TW1(T) should be classified as a novel species of the genus Paenibacillus, for which the name Paenibacillus xylaniclasticus sp. nov. is proposed. The type strain is TW1(T) (=NBRC 106381(T) =KCTC 13719(T) =TISTR 1914(T)).