EMAIL THIS PAGE TO A FRIEND
Journal of oncology pharmacy practice : official publication of the International Society of Oncology Pharmacy Practitioners
Determination of exposure of dispensary drug preparers to cyclophosphamide by passive sampling and liquid chromatography with tandem mass spectrometry.
PMID 22753612
Abstract
To determine cyclophosphamide exposure to preparers during tablet crushing and subsequent handling by analyzing indoor air collected using a high-performance volatile organic compounds-solvent desorption (VOC-SD) passive air sampler. The passive sampler was taped to the mask over the mouth of the preparer and indoor air was collected during crushing and preparation of cyclophosphamide tablets (Endoxan®). After collection, the carbon molecular sieve adsorbent of the passive sampler was placed in a centrifuge tube, and 1 mL of carbon disulfide was used to elute cyclophosphamide from the adsorbent. Liquid-liquid extraction with 1 mL of water was performed, and the aqueous phase was used as the test solution. Cyclophosphamide concentration was determined by liquid chromatography with ultraviolet and tandem mass spectrometry detection. Cyclophosphamide concentration was detected in the range of 7.6-157.7 ng/sampler. Our results showed that low-level exposure occurred near the mouth of the preparer, which could present risks for long-term exposure, especially if combined with multiple toxic drug exposures. The anticancer drug monitoring methodology described here is a simple exposure assessment that can be used to ensure the safety of hospital pharmacy tablet preparers. Furthermore, since the anticancer drug exposure risk is very high for preparers, preparation should be in hood or with face mask.