EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Chemical components of bee's pollen from buckwheat (Fagopyrum esculentum Moench)].
PMID 2275781
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74156
Nonacosane, analytical standard
C29H60
284246
Nonacosane, 99%
C29H60