EMAIL THIS PAGE TO A FRIEND
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Genistein stimulates jejunal chloride secretion via sex-dependent, estrogen receptor or adenylate cyclase mechanisms.
PMID 22759962
Abstract
Daily subcutaneous injections with the phytoestrogen genistein, 600 mg/ kg genistein/day (600G) significantly increased intestinal chloride (Cl(-)) secretion (I(sc), µA/cm(2)) in C57BL/6J female and male murine jejunum after 1-2-weeks treatment. In 600G females, basolateral application of the adenylate cyclase inhibitor MDL-12330A (10 µM) significantly reduced basal and total I(sc) in the presence of forskolin (27 and 40% respectively, P < 0.05), with no effect in 600G males, suggesting that 600G-mediated increases in I(sc) in females are due to an adenylate cyclase-dependent mechanism. Concomitant injections with the non-selective estrogen receptor (ER) antagonist ICI-182780 (25 mg/kg/day) resulted in a significant inhibition of basal I(sc) in males (38%, P < 0.05), but was without effect in females (further reinforcing an ER-independent mechanism of action). The ERα-selective antagonist (MPP, 25 mg/kg/day) similarly significantly inhibited the basal I(sc) (37%, P < 0.05) in males, whereas the ERβ-selective antagonist (PHTPP, 25 mg/kg/day) was without effect, suggesting that 600G-mediated increases in I (sc) in male mice are due to an ERα-dependent mechanism. Jejunum ERα/actin expression was significantly increased by 600G in males. Compared to intact mice, orchiectomy has differing effects on 600G-mediated basal Isc; castration (CAST) abolished the 600G-mediated increases in I(sc), and ovariectomy (OVX) had no effect on the 600G-stimulated increases in I(sc). Daily estradiol injections (10-20 mg/kg body weight estradiol (10E2 or 20E2) had no effect in intact females, whereas 10E2 significantly increased basal I(sc) in OVX females. These data suggest that daily estradiol and genistein injections have differential sex-dependent mechanisms of action on murine intestinal Cl(-) secretion.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M182
MDL-12,330A hydrochloride, ≥98% (HPLC), powder
C23H36N2 · HCl