EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Fenoterol functionally activates the β₃-adrenoceptor in human urinary bladder, comparison with rat and mouse: implications for drug discovery.
PMID 22760074
Abstract
Fenoterol has been reported to be a potent and selective β(2)-adrenoceptor agonist and is currently used clinically to treat asthma. Electrical field stimulation (EFS) of isolated urinary bladder mimics the voiding contraction by stimulating parasympathetic nerves, resulting in neurogenic contractions. To determine if stimulation of β(2)-adrenoceptors can inhibit this response, fenoterol was tested against EFS-induced contractions in human isolated urinary bladder and compared with mouse and rat. Bladder strips were mounted in organ baths and reproducible contractions induced by EFS. Fenoterol was added cumulatively in the presence of the β(2)-adrenoceptor antagonist ICI118551 or the β(3)-adrenoceptor antagonist L-748337. Fenoterol inhibited neurogenic contractions in all three species in a concentration-dependent manner with pEC(50) values of 6.66 ± 0.11, 6.86 ± 0.06 and 5.71 ± 0.1 in human, mouse and rat respectively. In human bladder strips ICI118551 (100 nM) did not affect responses to fenoterol, while L-748337 (0.3-3 μM) produced rightward shifts of the concentration-response curves with a pA(2) value of 8.10. In mouse bladder strips ICI118551 (30 nM) blocked the inhibitory effect of fenoterol (pA(2)=8.80), while L-748337 (10 μM) inhibited the response with a pA(2) of 5.79. In rat bladder ICI118551 (30 nM) was without effect, while L-748,337 (10 μM) inhibited the response to fenoterol with a pA(2) of 5.40. From these results it is clear that fenoterol potently activates β(3)-adrenoceptors in human isolated urinary bladder to inhibit EFS-induced contractions. Fenoterol also activates β(3)-adrenoceptors in rat, but β(2)-adrenoceptors in mouse bladder to inhibit EFS-induced contractions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F1016
Fenoterol hydrobromide
C17H21NO4 · HBr