EMAIL THIS PAGE TO A FRIEND
Deutsche medizinische Wochenschrift (1946)
[Supraventricular extrasystole after cardioversion with amiodarone].
PMID 22760409
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F6777
Flecainide acetate salt
C17H20F6N2O3 · C2H4O2