EMAIL THIS PAGE TO A FRIEND
The Pediatric infectious disease journal
Hyperacute infectious keratitis with Plesiomonas shigelloides following traumatic lamellar corneal laceration.
PMID 22760533
Abstract
Plesiomonas shigelloides rarely causes extraintestinal human disease, and infection of ocular tissues is even rarer, never having been reported as the sole pathogen of posttraumatic ocular infection. We report the first case of infectious keratitis due solely to P. shigelloides following traumatic corneal laceration and a literature review with regard to P. shigelloides ocular disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P2800000
Prednisolone acetate, European Pharmacopoeia (EP) Reference Standard
C23H30O6