EMAIL THIS PAGE TO A FRIEND
Journal of separation science
Microwave extraction and rapid isolation of arjunic acid from Terminalia arjuna (Roxb. ex DC.) stem bark and quantification of arjunic acid and arjunolic acid using HPLC-PDA technique.
PMID 22761141
Abstract
Arjunic acid and arjunolic acid are main bioactive components of Terminalia arjuna stem bark and reported for various biological activities. In this study, microwave-assisted extraction (MAE) of arjunic and arjunolic acid from stem bark of T. arjuna was investigated with developed and validated HPLC-PDA method, which resulted in the isolation of a novel anticancer molecule i.e. arjunic acid. Effects of several experimental parameters, such as type and volume of extraction solvents, microwave power, microwave extraction time, on the extraction efficiencies of arjunic, and arjunolic acid from stem bark of T. arjuna were evaluated. The optimal extraction conditions identified were 5.0 g quantity of stem bark powder, 20 mL of ethyl acetate, preleaching time 10 min, microwave power 600 W, temperature 65°C, and microwave irradiation time 5 min. The results showed that MAE is a more rapid extraction method with higher yield and lower solvent consumptions than reported methods. The HPLC-PDA analysis method was developed and validated to have good linearity, precision, sensitivity, and accuracy. MAE-HPLC-PDA is a faster, convenient, and appropriate method for isolation and determination of arjunic acid and arjunolic acid in the stem bark of T. arjuna.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00119
Arjunolic acid, ≥95% (LC/MS-ELSD)
C30H48O5