EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Coin exposure may cause allergic nickel dermatitis: a review.
PMID 22762130
Abstract
Nickel is used in coins because the metal has beneficial properties, including price, colour, weight, and corrosion resistance, and also because it is easy to stamp. It has often been claimed that the duration of skin contact with coins is too short to cause nickel release and dermatitis. However, it is well known by dermatologists specialized in occupational skin diseases, and by their nickel-allergic patients, that hand eczema in cashiers and other professionals who handle coins may be caused or aggravated by nickel release from coins. In this review, we present evidence from past studies showing that nickel-containing coins can indeed pose a risk for those who handle them. For protection of the health of consumers, cashiers, and other workers who handle coins, it is suggested that coins without nickel release should be used as a substitute for the high nickel-releasing coins currently in widespread use. The key risk factor in this situation is the ability of metal alloys in coins to release nickel and contaminate the skin after repeated contact from coin handling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
357553
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Ni
267007
Nickel, foil, thickness 0.25 mm, 99.995% trace metals basis
Ni
268259
Nickel, foil, thickness 0.125 mm, ≥99.9%
Ni
357588
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Ni
203904
Nickel, powder, <150 μm, 99.99% trace metals basis
Ni
266981
Nickel, powder, <50 μm, 99.7% trace metals basis
Ni
268283
Nickel, powder, <1 μm, 99.8% trace metals basis
Ni
577995
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Ni
267074
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Ni
267058
Nickel, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Ni
357626
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Ni
357634
Nickel, wire, diam. 0.25 mm, ≥99.9%
Ni
767484
Nickel, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
Ni
GF82156912
Nickel, rod, 100mm, diameter 12.0mm, 99%
Ni
GF11598194
Nickel, rod, 100mm, diameter 12.7mm, 99.98%
Ni
GF29576010
Nickel, rod, 100mm, diameter 16mm, 99.99+%
Ni
GF83584143
Nickel, rod, 100mm, diameter 2.0mm, 99.99+%
Ni
GF45193697
Nickel, rod, 100mm, diameter 2.0mm, annealed, 99%
Ni
GF11517948
Nickel, rod, 100mm, diameter 20.0mm, 99%
Ni
GF39205931
Nickel, rod, 100mm, diameter 25.4mm, 99.98%
Ni
GF69167920
Nickel, rod, 100mm, diameter 3.0mm, 99%
Ni
GF70332870
Nickel, rod, 100mm, diameter 3.3mm, 99.99+%
Ni
GF99799968
Nickel, rod, 100mm, diameter 30mm, 99%
Ni
GF90754241
Nickel, rod, 100mm, diameter 40mm, 99%
Ni
GF20561385
Nickel, rod, 100mm, diameter 5.0mm, 99.99+%
Ni
GF90932823
Nickel, rod, 100mm, diameter 6.35mm, 99%
Ni
GF50593002
Nickel, rod, 100mm, diameter 9.5mm, 99%
Ni
GF68355085
Nickel, rod, 200mm, diameter 1.5mm, 99.99+%
Ni
GF66250256
Nickel, rod, 200mm, diameter 10.0mm, 99.99+%
Ni
GF69613825
Nickel, rod, 200mm, diameter 12.0mm, 99%
Ni