EMAIL THIS PAGE TO A FRIEND
Journal of bioscience and bioengineering
Production of a new pyridine N-oxide by bioconversion with Cunninghamella echinulata var. elegans.
PMID 22762974
Abstract
A new N-oxide was produced from 3-(N-Boc-aminomethyl)-5-bromopyridine by bioconversion with Cunninghamella echinulata var. elegans ATCC 9245, and its structure was established based on the spectral data. The microbial N-oxidation is efficient and highly selective. The substrate was transformed into the product in 7 days.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
131652
Pyridine N-oxide, 95%
C5H5NO