EMAIL THIS PAGE TO A FRIEND
Environmental pollution (Barking, Essex : 1987)
Protozoan predation in soil slurries compromises determination of contaminant mineralization potential.
PMID 22763328
Abstract
Soil suspensions (slurries) are commonly used to estimate the potential of soil microbial communities to mineralize organic contaminants. The preparation of soil slurries disrupts soil structure, however, potentially affecting both the bacterial populations and their protozoan predators. We studied the importance of this "slurry effect" on mineralization of the herbicide 2-methyl-4-chlorophenoxyacetic acid (MCPA, (14)C-labelled), focussing on the effects of protozoan predation. Mineralization of MCPA was studied in "intact" soil and soil slurries differing in soil:water ratio, both in the presence and absence of the protozoan activity inhibitor cycloheximide. Protozoan predation inhibited mineralization in dense slurry of subsoil (soil:water ratio 1:3), but only in the most dilute slurry of topsoil (soil:water ratio 1:100). Our results demonstrate that protozoan predation in soil slurries may compromise quantification of contaminant mineralization potential, especially when the initial density of degrader bacteria is low and their growth is controlled by predation during the incubation period.