EMAIL THIS PAGE TO A FRIEND
The Journal of Neuroscience
Using motor behavior during an early critical period to restore skilled limb movement after damage to the corticospinal system during development.
PMID 22764234