EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and in vitro antitumor activity of novel scopoletin derivatives.
PMID 22765897
Abstract
Twenty scopoletin derivatives were developed by a systematic combinatorial chemical approach and their chemical structures were confirmed by MS, IR, (1)H NMR spectra and elemental analysis. Primary screening against mammary (MCF-7 and MDA-MB 231) and colon (HT-29) carcinoma cells indicated that five compounds (8d, 8g, 8j, 11b and 11g) displayed high antitumor potencies with IC(50) values below 20 μM whereas scopoletin showed IC(50) values above 100 μM. Moreover, the most promising compound 11g was more active than 5-fluorouracil. These results clearly indicated that the modification of the scopoletin structure could greatly increase its antitumor activity in vitro.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S2500
Scopoletin, ≥99%
C10H8O4
38332
Scopoletin, analytical standard
C10H8O4