EMAIL THIS PAGE TO A FRIEND
FEMS microbiology letters
Degradation of 3-chlorobiphenyl by in vivo constructed hybrid pseudomonads.
PMID 2276606
Abstract
3-Chlorobiphenyl-degrading bacteria were obtained from the mating between Pseudomonas putida strain BN10 and Pseudomonas sp. strain B13. Strains such as BN210 resulted from the transfer of the genes coding the enzyme sequence for the degradation of chlorocatechols from B13 into BN10, whereas B13 derivatives such as B131 have acquired the biphenyl degradation sequence from BN10. During growth of the hybrid strains on 3-chlorobiphenyl 90% chloride was released. Activities of phenylcatechol 2,3-dioxygenase, benzoate dioxygenase, catechol 1,2-dioxygenase, chloromuconate cyloisomerase and 4-carboxymethylenebut-2-en-4-olide hydrolase were found in 3-chlorobiphenyl-grown cells. The hybrid strains were found to convert some congeners of the Aroclor 1221 mixture such as mono- and dichloro-substituted biphenyls.