EMAIL THIS PAGE TO A FRIEND
Colloids and surfaces. B, Biointerfaces
Polymeric material for anti-biofouling.
PMID 22766280
Abstract
Copolymerization of methyl methacrylate (MMA) and acrylate terminated poly(ethylene oxide-co-ethylene carbonate) (PEOC) macromonomer yields graft copolymer with PEOC side chains. By use of quartz crystal microbalance with dissipation (QCM-D), we have investigated the adsorption of proteins on the surface constructed by the copolymer in seawater. It shows the polymeric surface exhibits protein resistance depending on the content of the side chains. Marine field tests show the protein resistant surface can effectively inhibit marine biofouling for more than 12 weeks.
Related Materials