EMAIL THIS PAGE TO A FRIEND
European journal of paediatric neurology : EJPN : official journal of the European Paediatric Neurology Society
Ohtahara syndrome or early-onset West syndrome? A case with overlapping features and favorable response to vigabatrin.
PMID 22766350
Abstract
The so called "severe neonatal epilepsies with suppression-burst pattern" include early infantile epileptic encephalopathy, and early myoclonic encephalopathy. Both syndromes are characterized by pharmacoresistant seizures that appear in the first weeks (up to the third month) of life, an electroencephalographic suppression-burst pattern, and a grim prognosis. Many patients later present with other forms of epileptic encephalopathies with difficult-to-treat seizures, such as West syndrome, and those who survive usually suffer from severe neurodevelopmental troubles. We here report the case of a patient who presented at our center with features consistent with a mixed form of these epileptic encephalopathies, and favorable neurodevelopmental evolution. To draw attention on the potentially favorable effect of vigabatrin in early-onset epileptic encephalopathies. Case study. In our patient, seizures immediately stopped upon initiation of vigabatrin treatment, and his development and neurological examination at one year are normal. Vigabatrin should be considered as an early treatment option in early-onset epileptic encephalopathies.