EMAIL THIS PAGE TO A FRIEND
Water science and technology : a journal of the International Association on Water Pollution Research
Ultrasonic-assisted cloud point extraction for determination of nickel in water samples by flame atomic absorption spectrometry.
PMID 22766868
Abstract
A novel method for the determination of nickel was established by ultrasonic-assisted cloud point extraction (UA-CPE) prior to flame atomic absorption spectrometry (FAAS) analysis. The nickel reacted with N,N'-bis(salicylidene)-1,2-ethanediamine (BSE) to form hydrophobic chelates, which were extracted into the micelles of alpha-[3,5-dimethyl-1-(2-methylpropyl)hexyl]-omega- poly(oxy-2-ethanediyl) (Tergitol TMN-6). Tergitol TMN-6 was used as green nonionic surfactant. BSE was synthesized and checked by nuclear magnetic resonance (NMR) spectra. The phase diagrams of the binary system, water-surfactant (Tergitol TMN-6), and the ternary systems, water-surfactant-salt, were determined. The effects of experimental conditions including pH of sample solution, concentration of chelating agent and surfactant, ultrasonic power, equilibration temperature and incubation time were evaluated in order to enhance sensitivity of the method. Under the optimal conditions, the calibration graph was linear in the range of 10-500 μg L(-1). The values obtained for the limit of detection and enrichment factor were 1.0 μg L(-1) and 30, respectively. The method was successfully applied to the analysis of nickel in water samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K4894
Kolliphor® P 188, solid
15759
Kolliphor® P 188
78973
Kolliphor® P 188 micro
62035
Kolliphor® P 407, oxyethylene 71.5-74.9 %
50246
Kolliphor® P 407 micro
P5556
Poloxamer 188 solution, 10%, sterile-filtered, BioReagent, suitable for insect cell culture
16758
Poloxamer 407, purified, non-ionic
435406
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,100
435414
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,900
435422
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,000
435430
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,800
435449
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,900
435457
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~4,400
435465
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~5,800
412325
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~8,400
542342
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~14,600
435473
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~2,000
435481
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~2,700
435503
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~3,300
07579
Synperonic® F 108, surfactant, non-ionic
86216
Synperonic® PE P105, surfactant
713538
Synperonic® PE/P84
NP9 TERGITOL, Type NP-9
NP40S TERGITOL solution, Type NP-40, 70% in H2O