EMAIL THIS PAGE TO A FRIEND
Journal of proteome research
Alteration of leaf metabolism in Bt-transgenic rice (Oryza sativa L.) and its wild type under insecticide stress.
PMID 22768924
Abstract
Insecticide is always used to control the damage from pests, while the potential influence on plants is rarely known. Time-course metabolic changes of wild and Bacillus thuringiensis (Bt) transgenic rice (Oryza sativa L.) plants after insecticide treatment were investigated by using gas chromatography-mass spectrometry (GC-MS). A combined statistical strategy of 2-way ANOVA and multivariate analyses (principal component analysis and hierarchal cluster analysis) was performed to find the stress-associated effects. The results reveal that a wide range of metabolites were dynamically varied in both varieties as a response to insecticide, in multiple metabolic pathways, such as biosynthesis and metabolism of amino acids, carbohydrates, fatty acids, TCA cycle, and the shikimate/phenylpropanoid pathway, and most of the changes were correlated with the exposure time and dependent on the variety. A set of stress defenses were activated, including phytohormone signaling pathway, antioxidant defense system, shikimate-mediated secondary metabolism, and so on. In particular, insecticide led to much stronger regulations of signaling molecules (salicylate and the precursor of jasmonate) and antioxidants (α-tocopherol and dehydroascorbate/ascorbate) in Bt-transgenic variety at the early stage. Our results demonstrated that the Bt-transgenic rice had a more acute and drastic response to insecticide stress than its non-transgenic counterpart in antioxidant system and signaling regulation.