EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Crystal structure of the rice branching enzyme I (BEI) in complex with maltopentaose.
PMID 22771800
Abstract
Starch branching enzyme (SBE) catalyzes the cleavage of α-1,4-linkages and the subsequent transfer of α-1,4 glucan to form an α-1,6 branch point in amylopectin. We determined the crystal structure of the rice branching enzyme I (BEI) in complex with maltopentaose at a resolution of 2.2Å. Maltopentaose bound to a hydrophobic pocket formed by the N-terminal helix, carbohydrate-binding module 48 (CBM48), and α-amylase domain. In addition, glucose moieties could be observed at molecular surfaces on the N-terminal helix (α2) and CBM48. Amino acid residues involved in the carbohydrate bindings are highly conserved in other SBEs, suggesting their generally conserved role in substrate binding for SBEs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8792
Cellopentaose, ≥80%
C30H52O26