EMAIL THIS PAGE TO A FRIEND
Journal of occupational and environmental medicine
Do workplace and home protective practices protect farm workers? Findings from the "For Healthy Kids" study.
PMID 22772953
Abstract
To assess associations of protective workplace and home practices with pesticide exposure levels. Using data from orchard workers in the Yakima Valley, Washington, we examined associations of workplace and home protective practices with (1) urinary metabolite concentrations of dimethylthiophosphate (DMTP) in adults and children aged 2 to 6 years and (2) azinphos-methyl levels in house and vehicle dust. Data were collected from 95 orchard workers and 94 children. Contrary to expectation, adult farm workers who wore boots or washed hands using hand sanitizer had higher concentrations of DMTP than those who did not. Children who attended daycare had higher DMTP concentrations than children who did not. Few workplace or home practices were associated with pesticide exposure levels; workers who used hand sanitizer had higher concentrations of DMTP, as did children who attended daycare.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45333
Azinphos-methyl, PESTANAL®, analytical standard
C10H12N3O3PS2