EMAIL THIS PAGE TO A FRIEND
In Vivo
Metallothionein Colon Crypt Immuno-positivity as a Rapid In Vivo Essay for Drug Efficacy Studies.
PMID 22773584
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-599
Anti-acetyl-Histone H3 Antibody, clone, from rabbit