EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
A photochemical route to 2-substituted benzo[b]furans.
PMID 22774767
Abstract
2-Substituted benzo[b]furans were synthesized by a one-step metal-free photochemical reaction between 2-chlorophenol derivatives and terminal alkynes by tandem formation of an aryl-C and a C-O bond via an aryl cation intermediate. The mild conditions and the application to chlorophenols rather of the more expensive bromo or iodo analogues makes this procedure environmentally convenient.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B8002
2,3-Benzofuran, 99%
C8H6O