EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Tandem ring-opening decarboxylation of cyclopropane hemimalonates with sodium azide: a short route to γ-aminobutyric acid esters.
PMID 22775774
Abstract
Cyclopropane hemimalonates, when treated with sodium azide, undergo a tandem ring-opening decarboxylation to produce γ-azidobutyric acids in good yields. These adducts were hydrogenated to form γ-aminobutyric acid (GABA) methyl esters.
Related Materials