EMAIL THIS PAGE TO A FRIEND
Archives of internal medicine
The consequences of ineffective regulation of dietary supplements.
PMID 22777632
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D167800
2,3-Dimethylmaleic anhydride, 98%
C6H6O3