EMAIL THIS PAGE TO A FRIEND
Chemistry, an Asian journal
Total synthesis of (±)-parvineostemonine.
PMID 22778054
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T89605
Tropinone, 99%
C8H13NO