EMAIL THIS PAGE TO A FRIEND
Molecular pharmaceutics
Mechanistic insights into PEPT1-mediated transport of a novel antiepileptic, NP-647.
PMID 22779445
Abstract
The present study, in general, is aimed to uncover the properties of the transport mechanism or mechanisms responsible for the uptake of NP-647 into Caco-2 cells and, in particular, to understand whether it is a substrate for the intestinal oligopeptide transporter, PEPT1 (SLC15A1). NP-647 showed a carrier-mediated, saturable transport with Michaelis-Menten parameters K(m) = 1.2 mM and V(max) = 2.2 μM/min. The effect of pH, sodium ion (Na(+)), glycylsarcosine and amoxicillin (substrates of PEPT1), and sodium azide (Na(+)/K(+)-ATPase inhibitor) on the flux rate of NP-647 was determined. Molecular docking and molecular dynamics simulation studies were carried out to investigate molecular interactions of NP-647 with transporter using homology model of human PEPT1. The permeability coefficient (P(appCaco-2)) of NP-647 (32.5 × 10(-6) cm/s) was found to be four times higher than that of TRH. Results indicate that NP-647 is transported into Caco-2 cells by means of a carrier-mediated, proton-dependent mechanism that is inhibited by Gly-Sar and amoxicillin. In turn, NP-647 also inhibits the uptake of Gly-Sar into Caco-2 cells and, together, this evidence suggests that PEPT1 is involved in the process. Docking and molecular dynamics simulation studies indicate high affinity of NP-647 toward PEPT1 binding site as compared to TRH. High permeability of NP-647 over TRH is attributed to its increased hydrophobicity which increases its affinity toward PEPT1 by interacting with the hydrophobic pocket of the transporter through hydrophobic forces.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G3127
Gly-Sar
C5H10N2O3
08591
Sodium azide 0.1 M solution