EMAIL THIS PAGE TO A FRIEND
Zhonghua jie he he hu xi za zhi = Zhonghua jiehe he huxi zazhi = Chinese journal of tuberculosis and respiratory diseases
[Advances in the research of rapid detection methods of pyrazinamide-resistant Mycobacterium tuberculosis].
PMID 22781157
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P4050000
Pyrazinamide, European Pharmacopoeia (EP) Reference Standard
C5H5N3O