EMAIL THIS PAGE TO A FRIEND
Annals of surgical oncology
A comparison of indocyanine green fluorescence imaging plus blue dye and blue dye alone for sentinel node navigation surgery in breast cancer patients.
PMID 22782671
Abstract
To evaluate two methods of sentinel node navigation surgery (SNNS) using blue dye with and without indocyanine green (ICG) fluorescence imaging (FI) to determine the usefulness of combined ICG and blue dye. Between 2005 and 2010, a total of 501 patients underwent SNNS in our hospital. Detection of sentinel lymph node (SLN) was performed with sulfan blue (SB) alone until 2008 and with a combination of SB and ICG-FI since 2009. ICG 5 mg and SB 15 mg were injected in the subareolar region, and FI was obtained by a fluorescence imaging device. We attempted to identify SLNs in 393 patients by SB alone and in 108 patients by a combination of SB and FI. The mean number of SLNs detected was 1.6 (0-5) for SB alone and 2.2 (1-6) for the combination method. The SLN identification rate was 95.7 % for SB alone and 100 % for the combination method so that the combination was significantly superior to SB in terms of the identification rate (p = 0.0037). In patients who received the combination method, detection of SLN was made through only SB in 1 patient, only ICG in 8 patients, and both in 99 patients. Lymph node metastasis was found in 56 patients with SB alone and in 16 patients with the combination method. Recurrence of an axillary node was observed in 3 patients (0.8 %) with SB alone and in no patients with the combination method. ICG-FI is a useful method and is especially recommended in cases where no radiotracers are available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
198218
Patent Blue VF, Dye content 50 %
C27H31N2NaO6S2
76357
Patent blue VF sodium salt, for microscopy
C27H31N2NaO6S2