EMAIL THIS PAGE TO A FRIEND
Journal of cutaneous medicine and surgery
Effective treatment of chronic hand dermatitis with 36 continuous months of alitretinoin administration: report of three cases.
PMID 22784520
Abstract
Moderate to severe chronic hand dermatitis (CHD) is not well controlled by current medical strategies; however, recent studies have shown significant improvement in patients treated with up to 6 months of oral alitretinoin (9-cis-retinoic acid). The results of longer-term continuous treatment are lacking. To evaluate the long-term safety and efficacy of alitretinoin for the treatment of CHD. The Physician's Global Assessment (PGA) and Modified Total Lesion Symptom Score (MTLSS) were used to assess CHD improvement in three patients treated with alitretinoin (10-30 mg/d). Patients had routine bloodwork and were monitored for adverse events. Significant improvement in MTLSS scores (≈ 76%) was seen at 2 months (p < .002) and maintained for the 3-year treatment period with occasional dosage adjustments. No significant adverse events developed that necessitated treatment withdrawal. Alitretinoin was well tolerated in the treatment of these three CHD patients who were carefully monitored over this prolonged treatment period.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R4643
9-cis-Retinoic acid, ≥98% (HPLC)
C20H28O2