EMAIL THIS PAGE TO A FRIEND
Cell calcium
PLCγ, G-protein of the Gαq type and cADPr pathway are associated to trigger the fertilization Ca2+ signal in the sea urchin egg.
PMID 22784667
Abstract
In all species, fertilization triggers in the egg a rapid and transient increase of intracellular free calcium (Cai), but how this signal is generated following sperm and egg interaction has not been clearly characterised yet. In sea urchin, a signalling pathway involving tyrosine kinase and PLCγ has been proposed to be at the origin of the fertilization Cai signal. We report here that injection of src homology-2 (SH2) domains of the sea urchin PLCγ inhibits in a competitive manner the endogenous PLCγ, alters both the amplitude and duration of the fertilization Cai wave, but does not abrogate it. Our results suggest that PLCγ acts in conjunction with a cADPr pathway and G-proteins of the Gαq type to trigger the fertilization Cai wave, and reinforce a crucial role for PLCγ at mitosis and cytokinesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C7344
Cyclic adenosine diphosphate-ribose, ≥90% (HPLC), lyophilized powder
C15H21N5O13P2