EMAIL THIS PAGE TO A FRIEND
Anti-cancer drugs
Tannic acid, an inhibitor of poly(ADP-ribose) glycohydrolase, sensitizes ovarian carcinoma cells to cisplatin.
PMID 22785358
Abstract
Tannic acid (TA) has been associated with anticancer functions in multiple tumor types both in vitro and in vivo. However, its effect on ovarian carcinoma cells has not been investigated, and its underlying anticancer mechanism(s) remain unclear. In this study, the effects of TA alone and in combination with cisplatin were evaluated using ovarian carcinoma cell lines. Combined treatment with TA and cisplatin was found to induce apoptosis and increase DNA damage in the cisplatin-resistant (SKOV-3 CDDP/R) and cisplatin-sensitive (SKOV-3) human ovarian carcinoma cell lines, respectively. TA was also found to enhance the toxicity of cisplatin in ovarian carcinoma cells associated with the inhibition of poly(ADP-ribose) glycohydrolase (PARG) expression, increase the accumulation of poly(ADP-ribose) (pADPr), following the release of apoptosis-inducing factor, and the activation of caspase-3. In conclusion, as a PARG inhibitor, TA showed anticancer activity and increased the sensitivity of SKOV-3 cells and SKOV-3 CDDP/R cell lines to cisplatin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
16201
Tannic acid, puriss., meets analytical specification of USP, powder
C76H52O46