EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Quantification of cyanamide in young seedlings of Vicia species, Lens culinaris, and Robinia pseudo-acacia by gas chromatography-mass spectrometry.
PMID 22785492
Abstract
We quantified the cyanamide content of young leaves of nine Vicia species, Lens culinaris, and Robinia pseudo-acacia using a modified analytical procedure that made it possible to measure the cyanamide content of a single leaf. Recent molecular phylogenetic analysis suggests that cyanamide is present in V. benghalensis, which is placed in a monophyletic group with cyanamide-biosynthesizing plants, V. villosa and V. cracca; this suggestion was verified.
Related Materials