EMAIL THIS PAGE TO A FRIEND
Circulation journal : official journal of the Japanese Circulation Society
Need and importance of biomarkers to predict acute myocardial infarction and its prognosis.
PMID 22786558