EMAIL THIS PAGE TO A FRIEND
Cancer discovery
SLC45A3-ELK4 chimera in prostate cancer: spotlight on cis-splicing.
PMID 22787087
Abstract
Using a series of detailed experiments, Zhang and colleagues establish that the prostate cancer RNA chimera SLC45A3-ELK4 is generated by cis-splicing between the 2 adjacent genes and does not involve DNA rearrangements or trans-splicing. The chimera expression is induced by androgen treatment likely by overcoming the read-through block imposed by the intergenic CCCTC insulators bound by CCCTC-binding factor repressor protein. The chimeric transcript, but not wild-type ELK4, is shown to augment prostate cancer cell proliferation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV100985
Anti-ELK4 antibody produced in rabbit, affinity isolated antibody