EMAIL THIS PAGE TO A FRIEND
Journal of toxicology and environmental health. Part A
Genetic polymorphisms in detoxification and DNA repair genes and susceptibility to glycidamide-induced DNA damage.
PMID 22788377
Abstract
Acrylamide (AA) is a probable human carcinogen formed in carbohydrate-rich foodstuffs upon heating. Glycidamide (GA), the AA metabolite formed by epoxidation, is considered the ultimate genotoxic agent. In this study, the in vitro genotoxic potential of AA and GA in human whole blood leukocytes was compared using the alkaline comet assay. Although AA did not induce significant DNA damage in the concentrations tested (up to 1000 μM), GA markedly increased the percentage of tail DNA at concentrations ≥250 μM. Further, this study addressed the role of genetic polymorphisms in key genes involved in metabolism and DNA repair pathways (BER, NER, HRR, and NHEJ) on GA-induced genotoxicity assessed by the alkaline comet assay. The results obtained suggested associations between DNA damage and polymorphisms of BER (MUTYH Gln335His and XRCC1 Gln399Arg) and NER (XPC Ala499Val) genes, either alone or in combination.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
04704
Glycidamide, analytical standard
C3H5NO2