EMAIL THIS PAGE TO A FRIEND
ACS applied materials & interfaces
Graphene from sugar and its application in water purification.
PMID 22788773
Abstract
This paper describes a green method for the synthesis of graphenic material from cane sugar, a common disaccharide. A suitable methodology was introduced to immobilize this material on sand without the need of any binder, resulting in a composite, referred to as graphene sand composite (GSC). Raman spectroscopy confirmed that the material is indeed graphenic in nature, having G and D bands at 1597 and 1338 cm(-1), respectively. It effectively removes contaminants from water. Here, we use rhodamine 6G (R6G) as a model dye and chloropyrifos (CP) as a model pesticide to demonstrate this application. The spectroscopic and microscopic analyses coupled with adsorption experiments revealed that physical adsorption plays a dominant role in the adsorption process. Isotherm data in batch experiments show an adsorption capacity of 55 mg/g for R6G and 48 mg/g for CP, which are superior to that of activated carbon. The adsorbent can be easily regenerated using a suitable eluent. This quick and cost-effective technique for the into a commercial water filter with appropriate engineering.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56226
Rhodamine 6G, analytical standard
C28H30N2O3 · HCl
252441
Rhodamine 6G perchlorate, Dye content 99 %
C28H30N2O3 · HClO4