EMAIL THIS PAGE TO A FRIEND
Progress in neuro-psychopharmacology & biological psychiatry
Effect of ACEA--a selective cannabinoid CB1 receptor agonist on the protective action of different antiepileptic drugs in the mouse pentylenetetrazole-induced seizure model.
PMID 22789660
Abstract
Endogenous cannabinoid ligands and cannabinoid CB1 receptor agonists have been shown to exert anticonvulsant effects in various experimental models of epilepsy. The purpose of this study was to determine the effects of arachidonyl-2'-chloroethylamide (ACEA-a highly selective cannabinoid CB1 receptor agonist) on the protective action of clonazepam, ethosuximide, phenobarbital, and valproate against pentylenetetrazole (PTZ)-induced clonic seizures in mice. To ascertain any pharmacokinetic contribution of ACEA to the observed interactions between tested drugs, free (non-protein bound) plasma and total brain concentrations of the antiepileptic drugs were estimated. Additionally, acute adverse-effect profiles of the combination of ACEA and different classical antiepileptic drugs (clonazepam, ethosuximide, phenobarbital and valproate) with respect to motor performance, long-term memory and skeletal muscular strength were measured. Results indicated that ACEA (10mg/kg, i.p.) co-administered with phenylmethylsulfonyl fluoride (PMSF-a substance protecting ACEA against degradation by the fatty-acid hydrolase; 30mg/kg, i.p.) significantly potentiated the anticonvulsant activity of ethosuximide, phenobarbital and valproate in the mouse PTZ-induced clonic seizure model by reducing their median effective doses (ED(50) values) from 122.8mg/kg to 71.7mg/kg (P<0.01; for ethosuximide), from 13.77mg/kg to 5.26mg/kg (P<0.05; for phenobarbital), and from 142.7mg/kg to 87.3mg/kg (P<0.05; for valproate), respectively. In contrast, ACEA (10mg/kg, i.p.) in combination with PMSF (30mg/kg, i.p.) had no impact on the protective action of clonazepam against PTZ-induced seizures in mice. However, ACEA (10mg/kg)+PMSF (30mg/kg) considerably increased free plasma and total brain concentrations of ethosuximide and valproate in mice suggesting a pharmacokinetic nature of interaction between drugs. In contrast, free plasma and total brain concentrations of clonazepam and phenobarbital remained unchanged after ACEA+PMSF administration and thus, indicating pharmacodynamic interactions. Moreover, none of the examined combinations of ACEA (10mg/kg, i.p.)+PMSF (30mg/kg, i.p.) with clonazepam, ethosuximide, phenobarbital, and valproate (at their ED(50) values from the PTZ-induced seizure test) affected motor coordination in the chimney test, long-term memory in the passive avoidance task, and muscular strength in the grip-strength test in mice, indicating no possible acute adverse effects in animals. In conclusion, pharmacodynamic enhancement of the anticonvulsant potency of phenobarbital by ACEA+PMSF is worthy of recommendation for further clinical settings. Pharmacokinetic interactions of ACEA+PMSF with ethosuximide and valproate seem to be responsible for a significant suppression of PTZ-induced seizures in mice. The combination of ACEA+PMSF with clonazepam seems to be neutral from a preclinical viewpoint.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
93482
Phenylmethanesulfonyl fluoride solution, ~0.1 M in ethanol (T)