EMAIL THIS PAGE TO A FRIEND
Current biology : CB
Plant cell biology: the ABC of monolignol transport.
PMID 22790004
Abstract
Lignins are complex aromatic heteropolymers that reinforce the cell walls of terrestrial plants. A new study identifies an ATP-binding cassette ABC transporter that pumps a monolignol lignin precursor across the plasma membrane.