EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Establishment of a monitoring system to detect inhibition of mRNA processing.
PMID 22790958
Abstract
A number of proteins complete mRNA processing in the nucleus, thus, inhibitor of mRNA processing is worth finding to analyze the mechanism of mRNA maturation in detail. Here, we established a monitoring system for mRNA processing using a test compound, spliceostatin A (SSA), which inhibits mRNA splicing. This system should serve to facilitate the discovery of novel compounds from natural resources that inhibit mRNA processing.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T5575
TG003, ≥98% (HPLC)
C13H15NO2S