EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Liquid chromatographic and thermal energy analyzer detection of N-nitrosodiphenylamine in formulations of diphenylamine.
PMID 2279134