PloS one
Plasma protein characteristics of long-term hemodialysis survivors.
PMID 22792249
Abstract
Hemodialysis (HD) patients are under recurrent circulatory stress, and hemodialysis has a high mortality rate. The characteristics of plasma proteomes in patients surviving long-term HD remain obscure, as well as the potential biomarkers in predicting prognoses. This study reports the proteome analyses of patient plasma from non-diabetic long-term HD (LHD, dialysis vintage 14.9±4.1 years, n = 6) and the age/sex/uremic etiology-comparable short-term HD (SHD, dialysis vintage 5.3±2.9 years, n = 6) using 2-DE and mass spectrometry. In addition, a 4-year longitudinal follow-up of 60 non-diabetic HD patients was subsequently conducted to analyze the baseline plasma proteins by ELISA in predicting prognosis. Compared to the SHD, the LHD survivors had increased plasma vitamin D binding proteins (DBP) and decreased clusterin, apolipoprotein A-IV, haptoglobin, hemopexin, complement factors B and H, and altered isoforms of α1-antitrypsin and fibrinogen gamma. During the 45.7±15 months for follow-up of the 60 HD patient cases, 16 patients died. Kaplan-Meier analysis demonstrated that HD patients with the lowest tertile of the baseline plasma DBP level have a significantly higher mortality rate. Multivariate Cox regression analysis further indicated that DBP is an independent predictor of mortality. In summary, the altered plasma proteins in LHD implicated accelerated atherosclerosis, defective antioxidative activity, increased inflammation/infection, and organ dysfunction. Furthermore, lower baseline plasma DBP in HD patients is related to mortality. The results suggest that the proteomic approach could help discover the potential biomarker in HD prognoses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
19899
CHAPS 100 mM solution
C32H58N2O7S