EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Triterpene compounds from Cirsium setosum].
PMID 22792796
Abstract
To investigate chemical constituents contained in cytotoxic petroleum ether extractive fractions from ethanol extracts of Cirsium setosum. The constituents were separated and purified by a combination of various chromatographic methods including silica gel, Sephadex LH-20, and preparative HPLC. Structures of the isolates were elucidated by spectroscopic methods including 1D, 2D NMR and MS methods. The compound structures were also determined by reference to literature. Twelve compounds were separated from the petroleum ether fraction of ethanolic extract and elucidated as lupenyl acetate (1), lupeol (2), lupenone (3), beta-amyrin (4), psi-taraxasterol (5), psi-taraxasteryl acetate (6), taraxasteryl acetate (7), marsformoxide B (8), alpha-amyrenone (9), beta-amyrenone (10), taraxasterone (11) and psi-taraxasterone (12). Of them, compounds 3, 5, 7-12 were separated from this genus for the first time.