EMAIL THIS PAGE TO A FRIEND
Advances in carbohydrate chemistry and biochemistry
Combining computational chemistry and crystallography for a better understanding of the structure of cellulose.
PMID 22794182