EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Molecular interaction fields based descriptors to interpret and compare chromatographic indexes.
PMID 22794793
Abstract
Molecular Interaction Fields (MIFs) based descriptors can be conveniently used to characterize and compare chromatographic scales. In this study, Quantitative Structure-Retention Relationships (QSRR) for eight different chromatographic systems were obtained with VolSurf+ descriptors and Partial Least Squares (PLS). A new and purpose-designed analysis tool highlights the different balance of intermolecular interactions governing solute retention, and estimates the similarity between chromatographic systems.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45324
Amitrol, PESTANAL®, analytical standard
C2H4N4