EMAIL THIS PAGE TO A FRIEND
ACS applied materials & interfaces
Supramolecular immobilization of xanthine oxidase on electropolymerized matrix of functionalized hybrid gold nanoparticles/single-walled carbon nanotubes for the preparation of electrochemical biosensors.
PMID 22801986
Abstract
Glassy carbon electrodes modified with single-walled carbon nanotubes and a three-dimensional network of electropolymerized Au nanoparticles capped with 2-mercaptoethanesulfonic acid, p-aminothiophenol, and 1-adamantanethiol were used as hybrid electrochemical platforms for supramolecular immobilization of a synthesized artificial neoglycoenzyme of xanthine oxidase and β-cyclodextrin through host-guest interactions. The ensemble was further employed for the bioelectrochemical determination of xanthine. The biosensor showed fast amperometric response within 5 s and a linear behavior in the 50 nM to 9.5 μM xanthine concentration range with high sensitivity, 2.47 A/(M cm(2)), and very low detection limit of 40 nM. The stability of the biosensor was significantly improved and the interferences caused by ascorbic and uric acids were noticeably minimized by coating the electrode surface with a Nafion thin film.